Eriodictyolchalcone ≥99% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

021282S5MG
100263-532EA 405.38 USD
100263-532
Eriodictyolchalcone ≥99% (by HPLC)
Eriodictyolchalcone
Formula: C₁₅H₁₂O₆
MDL Number: MFCD00017302
CAS Number: 14917-41-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/68/21285568.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR