2-Indolinone, solid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Oxindole2,3-Dihydro-2-indoloneIndolin-2-one2-Oxindole

01125625GM
100265-810EA 163.46 USD
100265-810
2-Indolinone, solid
2-Indolinone

  • Pale buff solid
  • Store in a cool, dry place

Formula: C₈H₇NO
MW: 133.15 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD00005711
CAS Number: 59-48-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/69/21285669.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR