1,2-Dioleoyl-3-stearoylglycerol ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

34182225MG
100290-538EA 555.77 USD
100290-538
1,2-Dioleoyl-3-stearoylglycerol ≥99%
1,2-Dioleoyl-3-stearoylglycerol
MDL Number: MFCD00673536
CAS Number: 113829-10-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/97/21287097.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR