1,2-Dioleoyl-3-palmitoylglycerol ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

34182125MG
100290-534EA 555.77 USD
100290-534
1,2-Dioleoyl-3-palmitoylglycerol ≥99%
1,2-Dioleoyl-3-palmitoylglycerol
Storage Temperature: Freezer
MDL Number: MFCD00272927
CAS Number: 65390-75-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/96/21287096.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR