n-Octacosane ≥99%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
N-octacosane
052800100MG
100277-530EA
255
USD
100277-530
n-Octacosane ≥99%
n-Octacosane
Formula:
H₃C(CH₂)₂₆CH₃ MW: 394.77 g/mol |
MDL Number:
MFCD00009355 CAS Number: 630-02-4 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...