1-Nonadecanol ≥99% for use in aqueous, textile, chemical and industrial foam control

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Nonadecyl alcohol

401900100MG
100298-774EA 197.46 USD
100298-774
1-Nonadecanol ≥99% for use in aqueous, textile, chemical and industrial foam control
1-Nonadecanol

For lipid research

Formula: H₃C(CH₂)₁₇CH₂OH
MW: 284.53 g/mol
MDL Number: MFCD00002824
CAS Number: 1454-84-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/71/24/21287124.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR