Z-L-Orn(Boc)-OH·DCHA ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

048295GM
100271-660EA 256.31 USD
100271-660
Z-L-Orn(Boc)-OH·DCHA ≥99%
Z-L-Orn(Boc)-OH·DCHA
Formula: C₁₈H₂₆N₂O₆·C₁₂H₂₃N
MDL Number: MFCD00270629
CAS Number: 13665-13-9
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR