Prunin ≥99% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

021160S10MG
100266-086EA 346.54 USD
100266-086
Prunin ≥99% (by HPLC), powder
Prunin

  • Cream colored powder
  • Store in a cool, dry place

Formula: C₂₁H₂₂O₁₀
Melting Pt: 222…224 °C
MDL Number: MFCD00210548
CAS Number: 529-55-5
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR