(±)-Naringenin ≥99% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Naringenin(±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one4',5,7-Trihydroxyflavanone5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one

021128S10MG
100269-782EA 137.31 USD
100269-782
(±)-Naringenin ≥99% (by HPLC), crystalline powder
(±)-Naringenin

  • Brown crystalline powder
  • Store in a cool, dry place

Formula: C₁₅H₁₂O₅
MW: 272.25 g/mol
Melting Pt: 254…256 °C
MDL Number: MFCD00006844
CAS Number: 480-41-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/59/27/21285927.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR