L-1-Naphthylalanine ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3-(1-Naphthyl)-L-alanineH-1-Nal-OH

0421591GM
100272-820EA 128.15 USD
100272-820
L-1-Naphthylalanine ≥99%, crystalline powder
L-1-Naphthylalanine

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₁₃H₁₃NO₂
MW: 215.252 g/mol
MDL Number: MFCD00066086
CAS Number: 55516-54-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/08/21286108.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR