1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥99%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione
, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione
26425325GM
100288-294EA
154.31
USD
100288-294
1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥99%
1-(2-Naphthoyl)-3,3,3-trifluoroacetone
Formula:
C₁₄H₉F₃O₂ MW: 266.22 g/mol Melting Pt: 74…76 °C |
ADR:
, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...