Monomyristin ≥99%

Synonyms: 2,3-Dihydroxypropyl tetradecanoate1-Monomyristoyl GlycerolGlycerol alpha-Monomyristate

311401250MG
100290-182EA 112.35 USD
100290-182
Monomyristin ≥99%
Monomyristin
Formula: C₁₇H₃₄O₄
MW: 302.46 g/mol
MDL Number: MFCD00046760
CAS Number: 589-68-4
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR