4-Methylumbelliferyl-Α-L-arabinopyranoside

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-Methylumbelliferyl alpha-L-Arabinopyranoside

M109025MG
100270-426EA 34 USD
100270-426
4-Methylumbelliferyl-Α-L-arabinopyranoside
4-Methylumbelliferyl-Α-L-arabinopyranoside
Formula: C₁₅H₁₆O₇
MW: 308.28 g/mol
MDL Number: MFCD00064087
CAS Number: 69414-26-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/59/59/21285959.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR