2-Methyl-5-(trifluoromethyl)benzonitrile ≥98%

0749625GM
100282-594EA 61.79 USD
100282-594
2-Methyl-5-(trifluoromethyl)benzonitrile ≥98%
2-Methyl-5-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃N
MDL Number: MFCD01631594
CAS Number: 261952-03-8
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/68/08/21286808.jpg https://us.vwr.com/stibo/web/std.lang.all/68/09/21286809.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR