Methyl tricosanoate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Tricosanoic acid methyl ester

202300100MG
100299-078EA 353.08 USD
100299-078
Methyl tricosanoate ≥99%
Methyl tricosanoate
Formula: C₂₄H₄₈O₂
MDL Number: MFCD00009349
CAS Number: 2433-97-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/75/40/21287540.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR