Methyl myristoleate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Myristoleic Acid Methyl EsterMethyl cis-9-tetradecenoate

201401100MG
100295-816EA 388 USD
100295-816
Methyl myristoleate ≥99%
Methyl myristoleate
Formula: C₁₅H₂₈O₂
MW: 240.387 g/mol
MDL Number: MFCD00056299
CAS Number: 56219-06-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/23/21287423.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR