Methyl-12-methyltetradecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

21141225MG
100274-924EA 562.31 USD
100274-924
Methyl-12-methyltetradecanoate ≥98%
Methyl-12-methyltetradecanoate
MDL Number: MFCD00053774
CAS Number: 5129-66-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/62/11/21286211.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR