Methyl-17-methyloctadecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

21181725MG
100274-950EA 623.77 USD
100274-950
Methyl-17-methyloctadecanoate ≥98%
Methyl-17-methyloctadecanoate
MDL Number: MFCD00083425
CAS Number: 55124-97-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/62/20/21286220.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR