Methyl-15-methylhexadecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

21161525MG
100274-938EA 623.77 USD
100274-938
Methyl-15-methylhexadecanoate ≥98%
Methyl-15-methylhexadecanoate
MDL Number: MFCD00083428
CAS Number: 6929-04-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/62/16/21286216.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR