Methyl-16-methylheptadecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

21171625MG
100274-946EA 623.77 USD
100274-946
Methyl-16-methylheptadecanoate ≥98%
Methyl-16-methylheptadecanoate
MDL Number: MFCD00072629
CAS Number: 5129-61-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/62/18/21286218.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR