Methyl-15-methylheptadecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

21171525MG
100274-944EA 623.77 USD
100274-944
Methyl-15-methylheptadecanoate ≥98%
Methyl-15-methylheptadecanoate
MDL Number: MFCD00214326
CAS Number: 54833-55-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/62/17/21286217.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR