Methyl-3-hydroxyhexanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Methyl 3-hydroxyhexanoate

24060325MG
100285-392EA 451.15 USD
100285-392
Methyl-3-hydroxyhexanoate ≥98%
Methyl 3-hydroxyhexanoate
Formula: C₇H₁₄O₃
MW: 146.1843 g/mol
MDL Number: MFCD00083583
CAS Number: 21188-58-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/69/54/21286954.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR