Methyl-2-fluorobenzoate ≥98%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
2-Fluorobenzoic acid emthyl ester
, Methyl 2-fluorobenzoate
09164525GM
100296-904EA
104.62
USD
100296-904
Methyl-2-fluorobenzoate ≥98%
Methyl 2-fluorobenzoate
Formula:
C₈H₇FO₂ MW: 154.14 g/mol |
MDL Number:
MFCD00017913 CAS Number: 394-35-4 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...