Methyl-3,4-difluorobenzoylformate

Supplier: INDOFINE CHEMICAL COMPANY MS

2613421GM
100293-746EA 138.62 USD
100293-746
Methyl-3,4-difluorobenzoylformate
Methyl 3,4-difluorobenzoylformate
Formula: C₁₀H₈F₂O₃
MDL Number: MFCD01631252
CAS Number: 220239-71-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/78/21287278.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR