Methyl 6-bromohexanoate ≥98%, liquid
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
6-Bromo-hexanoic acid methyl ester
196145GM
100267-612EA
45.77
USD
100267-612
Methyl 6-bromohexanoate ≥98%, liquid
Methyl 6-bromohexanoate
- Clear Colorless liquid
- Store in a cool, dry place
Formula:
C₇H₁₃BrO₂ MW: 209.084 g/mol |
ADR:
, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...