1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: a-D-Mannose pentaacetate

0241835GM
100263-052EA 180.46 USD
100263-052
1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose, powder
1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose

  • White powder
  • Store at 15-25 °C

Formula: C₁₆H₂₂O₁₁
MW: 390.341 g/mol
Melting Pt: 60…65.5 °C
MDL Number: MFCD00069798
CAS Number: 4163-65-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/34/21285534.jpg https://us.vwr.com/stibo/web/std.lang.all/55/35/21285535.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR