Primulin chloride, primuline yellow ≥95% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

020931S5MG
100268-624EA 451.15 USD
100268-624
Primulin chloride, primuline yellow ≥95% (by HPLC)
Primulin chloride, primuline yellow
Formula: C₂₃H₂₅O₁₂Cl
MDL Number: MFCD00185577
CAS Number: 30113-37-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/61/21285861.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR