D-Leucine benzyl ester-p-toluenesulfonate ≥99% (by TLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-D-Leu-OBzl.Tos-OHH-D-Leu-OBzl.TosOH

0419035GM
100270-502EA 137.31 USD
100270-502
D-Leucine benzyl ester-p-toluenesulfonate ≥99% (by TLC), powder
D-Leucine benzyl ester-p-toluenesulfonate

  • White powder
  • Store at 0-8 °C

Formula: C₁₃H₁₉NO₂
MW: 393.50 g/mol
Melting Pt: 131…147 °C
MDL Number: MFCD00066116
CAS Number: 17664-93-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/66/01/21286601.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR