Cyanidin-3-O-glucoside chloride ≥96% (by HPLC), powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
2-(3,4-Dihydroxyphenyl)-3-(β-D-glucopyranosyloxy)-5,7-dihydroxy-1-benzopyrylium Chloride
, Kuromanine Chloride
020915S5MG
100268-622EA
255
USD
100268-622
Cyanidin-3-O-glucoside chloride ≥96% (by HPLC), powder
Cyanidin-3-O-glucoside chloride
- Black-Red powder
- Store at -20 °C
Formula:
C₂₁H₂₁O₁₁Cl MW: 484.84 g/mol |
MDL Number:
MFCD00064923 CAS Number: 7084-24-4 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...