Apigenin-7-O-rutinoside ≥98.5% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

021121S10MG
100263-496EA 606.77 USD
100263-496
Apigenin-7-O-rutinoside ≥98.5% (by HPLC)
Apigenin-7-O-rutinoside
Formula: C₂₇H₃₀O₁₄
Melting Pt: 270…275 °C
MDL Number: MFCD00148889
CAS Number: 552-57-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/67/21285567.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR