Isopropyl 4,4,4-trifluoroacetoacetate ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4,4,4-Trifluoroacetoacetic Acid Isopropyl Ester

09159825GM
100298-038EA 179.15 USD
100298-038
Isopropyl 4,4,4-trifluoroacetoacetate ≥97%
Isopropyl 4,4,4-trifluoroacetoacetate
Formula: C₇H₉F₃O₃
MW: 198.14 g/mol
MDL Number: MFCD00040990
CAS Number: 175230-50-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/75/17/21287517.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR