6-O-a-D-Glucopyranosyl-D-glucitol ≥95%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Isomaltitol

0418771GM
100296-592EA 908.85 USD
100296-592
6-O-a-D-Glucopyranosyl-D-glucitol ≥95%
6-O-a-D-Glucopyranosyl-D-glucitol
Formula: C₁₂H₂₄O₁₁
MW: 344.30 g/mol
MDL Number: MFCD00083636
CAS Number: 534-73-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/41/21287441.jpg https://us.vwr.com/stibo/web/std.lang.all/74/42/21287442.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR