5-Iodoanthranilic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Amino-5-iodobenzoic acid

0196110GM
100268-494EA 36.62 USD
100268-494
5-Iodoanthranilic acid
5-Iodoanthranilic acid
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Melting Pt: ∼220 °C
MDL Number: MFCD00007849
CAS Number: 5326-47-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/47/21285847.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR