Cyanidin-3-O-galactoside chloride ≥97% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

020923S5MG
100269-758EA 490.38 USD
100269-758
Cyanidin-3-O-galactoside chloride ≥97% (by HPLC), powder
Cyanidin-3-O-galactoside chloride

  • Black red powder
  • Store at -20 °C, To be kept away from light and moisture

Formula: C₂₁H₂₁O₁₁Cl
MW: 484.85 g/mol
MDL Number: MFCD00270387
CAS Number: 27661-36-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/59/20/21285920.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR