Hypericin ≥98% (by HPLC), crystals

020468S10MG
100263-404EA 347.75 USD
100263-404
Hypericin ≥98% (by HPLC), crystals
Hypericin

  • Blue Black crystals
  • To be kept in a desiccator

Formula: C₃₀H₁₆O₈
Melting Pt: 320 °C
MDL Number: MFCD00016683
CAS Number: 548-04-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/56/21285556.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR