6-(Trifluoromethyl)-4-quinolinol ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-Hydroxy-6-(trifluoromethyl)quinoline

271701GM
100289-636EA 90.15 USD
100289-636
6-(Trifluoromethyl)-4-quinolinol ≥97%
6-(Trifluoromethyl)-4-quinolinol
Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
Melting Pt: 265…268 °C
MDL Number: MFCD00209673
CAS Number: 49713-51-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/54/21287054.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR