7-Hydroxy-4-(trifluoromethyl)coumarin ≥98%, crystalline solid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: TFMU

15006250MG
100263-864EA 37.92 USD
100263-864
7-Hydroxy-4-(trifluoromethyl)coumarin ≥98%, crystalline solid
7-Hydroxy-4-(trifluoromethyl)coumarin

  • Pinkish beige crystal solid
  • Store in a cool, dry place

Formula: C₁₀H₅F₃O₃
MW: 230.14 g/mol
Melting Pt: 178…180 °C
MDL Number: MFCD00037578
CAS Number: 575-03-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/86/21285586.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR