4-Hydroxybenzylacetone ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-(4-Hydroxyphenyl)-2-butanone1-(4-Hydroxyphenyl)-3-butanone

02882525GM
100263-856EA 170 USD
100263-856
4-Hydroxybenzylacetone ≥98%
4-Hydroxybenzylacetone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Melting Pt: 80…84 °C
MDL Number: MFCD00002394
CAS Number: 5471-51-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/85/21285585.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR