4-Hydroxy-6-methylcoumarin ≥98%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

0205665GM
100265-956EA 78.46 USD
100265-956
4-Hydroxy-6-methylcoumarin ≥98%, Powder
4-Hydroxy-6-methylcoumarin

  • White powder
  • Store in a cool, dry place

Formula: C₁₀H₈O₃
Melting Pt: 262…264 °C
MDL Number: MFCD00085738
CAS Number: 13252-83-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/85/21285685.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR