Methyl chrysin,Tectochrysin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

H11050MG
100265-726EA 85 USD
100265-726
Methyl chrysin,Tectochrysin ≥98%, powder
Methyl chrysin,Tectochrysin

  • Yellow powder
  • Store in a cool, dry place

Formula: C₁₆H₁₂O₄
Melting Pt: 166…168 °C
MDL Number: MFCD00016945
CAS Number: 520-28-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/63/21285663.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR