4-Hydroxy-6-methoxycoumarin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-Hydroxy-6-methoxy-2H-chromen-2-one

19348250MG
100267-192EA 26.15 USD
100267-192
4-Hydroxy-6-methoxycoumarin ≥98%, powder
4-Hydroxy-6-methoxycoumarin

  • Off White powder
  • Store in a cool, dry place

Formula: C₁₀H₈O₄
MW: 192.17 g/mol
Melting Pt: 268…270 °C
MDL Number: MFCD00463526
CAS Number: 13252-84-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/44/21285744.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR