5'-Hydroxy-2'-methoxyacetophenone ≥98%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-(5-Hydroxy-2-methoxyphenyl)ethanone

195941GM
100269-472EA 94.05 USD
100269-472
5'-Hydroxy-2'-methoxyacetophenone ≥98%, Powder
5'-Hydroxy-2'-methoxyacetophenone

  • Pale cream colored powder
  • Store in a cool, dry place

Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
Melting Pt: 82,2…84.7 °C
MDL Number: MFCD04038299
CAS Number: 31405-60-4
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR