3-Hydroxyhexadecanoic acid ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

14160350MG
100295-796EA 614.62 USD
100295-796
3-Hydroxyhexadecanoic acid ≥98%
3-Hydroxyhexadecanoic acid
MDL Number: MFCD00171638
CAS Number: 2398-34-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/21/21287421.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR