7-Hydroxycoumarin ≥99%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Umbelliferone7-Hydroxy-2-benzopyrone7-Hydroxy-2H-chromen-2-one

H0175GM
100265-716EA 28.77 USD
100265-716
7-Hydroxycoumarin ≥99%, Powder
7-Hydroxycoumarin

  • Off white powder
  • Store in a cool, dry place

Formula: C₉H₆O₃
MW: 162.14 g/mol
Melting Pt: 231…233 °C (dec.)
MDL Number: MFCD00006878
CAS Number: 93-35-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/62/21285662.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR