2-Hydroxychalcone ≥98%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3-(2-Hydroxyphenyl)-1-phenylprop-2-en-1-one2-Hydroxybenzylidenacetophenone

0212705GM
100266-104EA 78.46 USD
100266-104
2-Hydroxychalcone ≥98%, crystalline powder
2-Hydroxychalcone

  • Fine yellow crystalline powder
  • Store in a cool, dry place

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: 150…153 °C
MDL Number: MFCD00016449
CAS Number: 644-78-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/03/21285703.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR