4',5,7-Trihydroxy-3'-methoxyflavanone ≥90% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

02009150MG
100268-506EA 717.92 USD
100268-506
4',5,7-Trihydroxy-3'-methoxyflavanone ≥90% (by HPLC), powder
4',5,7-Trihydroxy-3'-methoxyflavanone

  • Cream powder
  • 0

Formula: C₁₆H₁₄O₆
Melting Pt: 223…225 °C
MDL Number: MFCD00017314
CAS Number: 107657-60-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/48/21285848.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR