Caproic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Hexanoic acid

100600100MG
100293-552EA 88.92 USD
100293-552
Caproic acid ≥99%
Caproic acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00004421
CAS Number: 142-62-1
UN: 2829
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/72/60/21287260.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR