n-Hexane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: HexanenHexaneHexyl hydride

050600100MG
100274-318EA 107.23 USD
100274-318
n-Hexane ≥99%
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/61/83/21286183.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR