10(Z)-Heptadecenoic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (Z)-Heptadec-10-enoic acid

101701100MG
100293-564EA 333.46 USD
100293-564
10(Z)-Heptadecenoic acid ≥99%
10(Z)-Heptadecenoic acid
Formula: C₁₇H₃₂O₂
MW: 268.4348 g/mol
MDL Number: MFCD00133175
CAS Number: 29743-97-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/63/21287263.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR