n-Heneicosane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

052100100MG
100277-524EA 268.08 USD
100277-524
n-Heneicosane ≥99%
n-Heneicosane
Formula: H₃C(CH₂)₁₉CH₃
MW: 296.58 g/mol
MDL Number: MFCD00009346
CAS Number: 629-94-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/54/21286454.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR